EMAIL THIS PAGE TO A FRIEND
Military medicine
Dehydroepiandrosterone and dehydroepiandrosterone sulfate: anabolic, neuroprotective, and neuroexcitatory properties in military men.
PMID 23356127
Abstract
Evidence links dehydroepiandrosterone (DHEA) and dehydroepiandrosterone sulfate (DHEAS) to crucial military health issues, including operational stress, resilience, and traumatic brain injury. This study evaluated the anabolic, neuroprotective, and neuroexcitatory properties of DHEA(S) in healthy military men. A salivary sample was obtained from 42 men and assayed for DHEA(S), testosterone, nerve growth factor (NGF; which supports nerve cell proliferation), and salivary alpha amylase (sAA; a proxy of sympathetic nervous system function). Separate regression analyses were conducted with DHEA and DHEAS as independent variables, and testosterone, NGF, and sAA as dependent variables, respectively. The models explained 23.4% of variance in testosterone (p < 0.01), 17.2% of variance in NGF (p < 0.01), and 7.4% of variance in sAA (p = 0.09). Standardized beta coefficients revealed that DHEA independently influenced testosterone (beta = 0.40, p < 0.01), whereas DHEAS independently influenced NGF (beta = 0.48, p < 0.01) and sAA (beta = 0.36, p < 0.05). DHEA demonstrated anabolic properties, whereas DHEAS demonstrated neuroprotective and neuroexcitatory properties in military men. This area of study has broad implications for stress inoculation, traumatic brain injury rehabilitation, and regenerative medicine in military personnel.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8220
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
10065 α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
86250 α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
A9857
α-Amylase from Aspergillus oryzae, powder, ≥150 units/mg protein (biuret)
A7595
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
A3403
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
A4582 α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
A4551
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
A4862 α-Amylase from Bacillus licheniformis, liquid
A6814 α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
A6380 α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
10069 α-Amylase from Bacillus subtilis, powder, yellow-brown, ~380 U/mg
10070 α-Amylase from Bacillus subtilis, powder, yellow-brown, ~50 U/mg
A9972
α-Amylase from human pancreas, lyophilized powder, ≥100 units/mg protein
A1031
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
A0521
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
A6255
α-Amylase from porcine pancreas, DFP Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
A4268
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
D-063
Dehydroepiandrosterone (DHEA) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
C19H28O2
86247 Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg