EMAIL THIS PAGE TO A FRIEND
Biochemical Society transactions
Palmitoylation, pathogens and their host.
PMID 23356263
Abstract
S-Palmitoylation, the only reversible post-translational lipid modification, confers unique biochemical and functional properties to proteins. Although it has long been known that viral proteins are palmitoylated, recent studies reveal that this modification plays a critical role for pathogens of all kinds and at multiple steps of their life cycle. The present review examines the involvement of S-palmitoylation in infection by viruses, bacteria and parasites and illustrates how pathogens have evolved to manipulate the host palmitoylation machinery.