EMAIL THIS PAGE TO A FRIEND
The FEBS journal
QM/MM modelling of ketosteroid isomerase reactivity indicates that active site closure is integral to catalysis.
PMID 23356661
Abstract
Ketosteroid isomerase (Δ⁵-3-keto steroid isomerase or steroid Δ-isomerase) is a highly efficient enzyme at the centre of current debates on enzyme catalysis. We have modelled the reaction mechanism of the isomerization of 3-oxo-Δ⁵-steroids into their Δ⁴-conjugated isomers using high-level combined quantum mechanics/molecular mechanics (QM/MM) methods, and semi-empirical QM/MM molecular dynamics simulations. Energy profiles were obtained at various levels of QM theory (AM1, B3LYP and SCS-MP2). The high-level QM/MM profile is consistent with experimental data. QM/MM dynamics simulations indicate that active site closure and desolvation of the catalytic Asp38 occur before or during formation of dienolate intermediates. These changes have a significant effect on the reaction barrier. A low barrier to reaction is found only when the active site is closed, poising it for catalysis. This conformational change is thus integral to the whole process. The effects on the barrier are apparently largely due to changes in solvation. The combination of high-level QM/MM energy profiles and QM/MM dynamics simulation shows that the reaction involves active site closure, desolvation of the catalytic base, efficient isomerization and re-opening of the active site. These changes highlight the transition between the ligand binding/releasing form and the catalytic form of the enzyme. The results demonstrate that electrostatic interactions (as a consequence of pre-organization of the active site) are crucial for stabilization during the chemical reaction step, but closure of the active site is essential for efficient catalysis to occur.
Related Materials