EMAIL THIS PAGE TO A FRIEND
Meat science
Sensory evaluation of boar loins: trained assessors' olfactory acuity affects the perception of boar taint compounds.
PMID 23357575
Abstract
This study investigated the impact of assessors' varying olfactory acuity on the perceived intensity of androstenone and skatole odour and flavour in boar loins. To discriminate sensitive (SENS) and highly sensitive (SENSHIGH) panellists, two levels of androstenone were used on smell strips. Sensitivity was defined as the correct identification of the androstenone strip in three replicate triangle tests. Judges then assessed loins from boars, castrated pigs and gilts. SENSHIGH assessors scored low-fat boar loins with 1.5 to 2.0μg of androstenone per gram of melted back fat which is significantly different from castrate and gilt loins for androstenone odour and flavour whereas SENS assessors were less discriminating. Panellists' olfactory acuity should thus be considered for selection and training. The presented paper strip system is suggested for objective screening and training purposes and to be used as quantitative references in descriptive analysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M51458
3-Methylindole, 98%
C9H9N
W301912
Skatole, ≥98%
C9H9N