EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Solvent dependent reductive defluorination of aliphatic C-F bonds employing Sm(HMDS)2.
PMID 23358653
Abstract
Sm(HMDS)(2) in n-hexane mediates fast cleavage of primary, secondary and tertiary alkyl fluorides in good to excellent yields. In n-hexane Sm(HMDS)(2) exhibits uniquely enhanced reductive ability towards the C-F bond compared to when using THF as solvent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
324620
Lithium bis(trimethylsilyl)amide, 97%
C6H18LiNSi2
225770
Lithium bis(trimethylsilyl)amide solution, 1.0 M in THF
C6H18LiNSi2
577928
Lithium bis(trimethylsilyl)amide solution, 1 M in toluene
C6H18LiNSi2
766917
Lithium bis(trimethylsilyl)amide solution, 1.5 M in THF
C6H18LiNSi2
224367
Lithium bis(trimethylsilyl)amide solution, 1.0 M in hexanes
C6H18LiNSi2
775053
Lithium bis(trimethylsilyl)amide solution, 0.5 M in 2-methyltetrahydrofuran
C6H18LiNSi2
771864
Lithium bis(trimethylsilyl)amide solution, 0.5 M in toluene
C6H18LiNSi2
577014
Lithium bis(trimethylsilyl)amide solution, 1 M in tert-butyl methyl ether
C6H18LiNSi2
324671
Potassium bis(trimethylsilyl)amide, 95%
C6H18KNSi2
702722
Potassium bis(trimethylsilyl)amide solution, 1 M in THF
C6H18KNSi2
277304
Potassium bis(trimethylsilyl)amide solution, 0.5 M in toluene
C6H18KNSi2
775061
Potassium bis(trimethylsilyl)amide solution, 1.0 M in 2-methyltetrahydrofuran
C6H18KNSi2
766941
Potassium bis(trimethylsilyl)amide solution, 1.0 M in methyl tert-butyl ether
C6H18KNSi2
261203
Samarium, −40 mesh, 99% trace rare earth metals basis
Sm
263184
Samarium, ingot, 99.9% trace rare earth metals basis
Sm
693731
Samarium, foil, thickness 1 mm, 99.9% trace rare earth metals basis
Sm
235083
Sodium bis(trimethylsilyl)amide, 95%
C6H18NNaSi2
245585
Sodium bis(trimethylsilyl)amide solution, 1.0 M in THF
C6H18NNaSi2
80631
Sodium bis(trimethylsilyl)amide solution, produced by Wacker Chemie AG, Burghausen, Germany, 40% in THF
C6H18NNaSi2
368059
Sodium bis(trimethylsilyl)amide solution, 0.6 M in toluene
C6H18NNaSi2
771546
Sodium bis(trimethylsilyl)amide solution, 0.5 M in methyl tert-butyl ether
C6H18NNaSi2
547816
Tris[N,N-bis(trimethylsilyl)amide]europium(III), 98%
C18H54EuN3Si6
547824
Tris[N,N-Bis(trimethylsilyl)amide]gadolinium(III), 98%
C18H54GdN3Si6
547980
Tris[N,N-bis(trimethylsilyl)amide]samarium(III), 98%
C18H54N3Si6Sm
524514
Tris[N,N-bis(trimethylsilyl)amide]yttrium
C18H54N3Si6Y
702021
Tris[N,N-bis(trimethylsilyl)amide]yttrium, packaged for use in deposition systems
C18H54N3Si6Y