EMAIL THIS PAGE TO A FRIEND
Journal of clinical oncology : official journal of the American Society of Clinical Oncology
Cytochrome P17 inhibition with ketoconazole as treatment for advanced granulosa cell ovarian tumor.
PMID 23358981