EMAIL THIS PAGE TO A FRIEND
Nature biotechnology
Efficient genome editing in zebrafish using a CRISPR-Cas system.
PMID 23360964
Abstract
In bacteria, foreign nucleic acids are silenced by clustered, regularly interspaced, short palindromic repeats (CRISPR)--CRISPR-associated (Cas) systems. Bacterial type II CRISPR systems have been adapted to create guide RNAs that direct site-specific DNA cleavage by the Cas9 endonuclease in cultured cells. Here we show that the CRISPR-Cas system functions in vivo to induce targeted genetic modifications in zebrafish embryos with efficiencies similar to those obtained using zinc finger nucleases and transcription activator-like effector nucleases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CRISPR Custom CRISPR Products