EMAIL THIS PAGE TO A FRIEND
Journal of analytical toxicology
Analysis of synthetic cathinones commonly found in bath salts in human performance and postmortem toxicology: method development, drug distribution and interpretation of results.
PMID 23361867
Abstract
To date, the Toxicology Section of the Montgomery County Coroner's Office/Miami Valley Regional Crime Laboratory has identified six synthetic cathinones, commonly found in bath salt products, in 43 cases. Thirty-two cases will be reviewed here, including all of the postmortem cases, all of the human performance cases that had blood specimens submitted, and one urine-only human performance case. The following compounds have been confirmed: 3,4-methylenedioxypyrovalerone (MDPV), 3,4-methylenedioxymethcathinone (methylone), pyrovalerone, pentylone, alpha-pyrrolidinopentiophenone (alpha-PVP) and methedrone. The method also screens for mephedrone, butylone and 3-fluoromethcathinone. Case demographics show 42 white males and females ranging in age from 19 to 53 years. The remaining case was that of a 34-year-old Hispanic male. The 43 cases represent 17 driving under the influence, two domestic violence, four suicides, 12 overdoses, six accidents, one drug-facilitated assault and one homicide. Data will be presented on the distribution of some of these cathinones in various matrices. After review, blood concentration does not appear to predict outcome regarding fatalities or impairment. The highest MDPV concentration occurred in a suicide by hanging and the highest methylone concentration was in a driver. The confirmation method is a liquid-liquid extraction with detection by liquid chromatography triple quadrupole mass spectrometry using electrospray ionization in multiple reaction monitoring mode.