EMAIL THIS PAGE TO A FRIEND
Zhonghua xue ye xue za zhi = Zhonghua xueyexue zazhi
[Thrombosis of the cerebral veins and sinuses in acute promyelocytic leukemia after all-trans retinoic acid treatment: a case report and literature review].
PMID 23363754