EMAIL THIS PAGE TO A FRIEND
Journal of clinical microbiology
Invasive pulmonary aspergillosis due to Emericella nidulans var. echinulata, successfully cured by voriconazole and micafungin.
PMID 23363828
Abstract
A 78-year-old male who was undergoing prolonged glucocorticoid treatment experienced cough and expectoration for 2 weeks. Galactomannan antigen analysis and a chest computed tomography (CT) scan suggested a diagnosis of invasive pulmonary aspergillosis. DNA sequencing indicated that Emericella nidulans var. echinulata was the causative agent. A combination of voriconazole and micafungin successfully treated the illness.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48230 D-Galacto-D-mannan from Ceratonia siliqua, ~95% (HPLC)