EMAIL THIS PAGE TO A FRIEND
Chest
Inhaled corticosteroid dose response using domiciliary exhaled nitric oxide in persistent asthma: the FENOtype trial.
PMID 23364390
Abstract
International guidelines advocate a standard approach to asthma management for all, despite its heterogeneity. "Personalized" treatment of inflammatory asthma phenotypes confers superior benefits. We wished to evaluate dose response to inhaled corticosteroids (ICSs) in patients with asthma with an elevated fractional exhaled nitric oxide (Feno) phenotype using domiciliary measurements. We performed a randomized, crossover trial in 21 patients with mild to moderate persistent asthma receiving ICSs with elevated Feno (>30 parts per billion [ppb]) that increased further (>10 ppb) after ICS washout. Patients were randomized to 2 weeks of either fluticasone propionate 50 μg bid (FP100) or 250 μg bid (FP500). The primary outcome was response in diurnal domiciliary Feno levels. Secondary outcomes included mannitol challenge, serum eosinophilic cationic protein (ECP), blood eosinophil count, and asthma control questionnaire. We found significant dose-related reductions of diurnal Feno compared with baseline - morning Feno: baseline = 71 ppb (95% CI, 61-83 ppb); FP100 = 34 ppb (95% CI, 29-40 ppb), P < .001; FP500 = 27 ppb (95% CI, 22-33 ppb), P < .001; and significant dose separation for morning, P < .05, and evening, P < .001. Time-series Feno displayed exponential decay: FP100 R² = 0.913, half-life = 69 h (95% CI, 50-114 h); FP500 R² = 0.966, half-life = 55 h (95% CI, 45-69 h), as well as diurnal variation. The Asthma Control Questionnaire showed significant improvements exceeding the minimal important difference (>0.5) with values in keeping with controlled asthma (<0.75) after each dose: FP100 = 0.48 (95% CI, 0.24-0.71), P = .004; FP500 = 0.37 (95% CI, 0.18-0.57), P = .001. All other secondary inflammatory related outcomes (mannitol, ECP, and eosinophils) showed significant improvements from baseline but no dose separation. There is a significant dose response of diurnal Feno to ICS in patients with asthma with an elevated Feno phenotype, which translates into well-controlled asthma. Further interventional studies are warranted using domiciliary Feno in this specific phenotype. ClinicalTrials.gov; No.: NCT00995657; URL: clinicaltrials.gov.