EMAIL THIS PAGE TO A FRIEND
Journal of clinical rheumatology : practical reports on rheumatic & musculoskeletal diseases
Primary meningococcal arthritis requiring surgical drainage.
PMID 23364663
Abstract
Primary meningococcal arthritis (PMA) is a relatively rare diagnosis where the role of early surgical intervention for its treatment is not well defined. We report a case of PMA in a young otherwise healthy patient who developed polyarticular joint pain secondary to Niessieria meningitidis without systemic symptoms of meningitis or meningococcemia. He underwent a prolonged course of intravenous antibiotics and serial aspirations of his shoulder. However, symptoms in his shoulder did not improve and he later underwent surgical irrigation and debridement.Intraoperatively, the patient had no signs of articular damage to his right shoulder despite prolonged clinically symptomatic disease. Six weeks after surgery, he has regained normal strength and full range of motion without any deficits.Nonoperative management of PMA is frequently, but not invariably, successful. We report a patient with this diagnosis who ultimately needed surgical evacuation of his shoulder joint to achieve resolution of his symptoms.
Related Materials