EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Multisite binding of a general anesthetic to the prokaryotic pentameric Erwinia chrysanthemi ligand-gated ion channel (ELIC).
PMID 23364792
Abstract
Pentameric ligand-gated ion channels (pLGICs), such as nicotinic acetylcholine, glycine, γ-aminobutyric acid GABA(A/C) receptors, and the Gloeobacter violaceus ligand-gated ion channel (GLIC), are receptors that contain multiple allosteric binding sites for a variety of therapeutics, including general anesthetics. Here, we report the x-ray crystal structure of the Erwinia chrysanthemi ligand-gated ion channel (ELIC) in complex with a derivative of chloroform, which reveals important features of anesthetic recognition, involving multiple binding at three different sites. One site is located in the channel pore and equates with a noncompetitive inhibitor site found in many pLGICs. A second transmembrane site is novel and is located in the lower part of the transmembrane domain, at an interface formed between adjacent subunits. A third site is also novel and is located in the extracellular domain in a hydrophobic pocket between the β7-β10 strands. Together, these results extend our understanding of pLGIC modulation and reveal several specific binding interactions that may contribute to modulator recognition, further substantiating a multisite model of allosteric modulation in this family of ion channels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36972
Bromoform, amylene stabilized, analytical standard
CHBr3
241032
Bromoform, contains 60-120 ppm 2-methyl-2-butene as stabilizer, 99%
CHBr3
132942
Bromoform, contains 1-3% ethanol as stabilizer, 96%
CHBr3
40212 Bromoform solution, certified reference material, 5000 μg/mL in methanol
CHBr3
C2432
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
CHCl3
650498
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
CHCl3
650471
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
CHCl3
366927
Chloroform, for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
CHCl3
34854
Chloroform, for HPLC, ≥99.8%, amylene stabilized
CHCl3
496189
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
CHCl3
366919
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
CHCl3
472476
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
CHCl3
319988
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
CHCl3
132950
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
CHCl3
32211
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
CHCl3
24216
Chloroform, contains ethanol as stabilizer, meets analytical specification of DAB9, BP, 99-99.4% (GC)
CHCl3