EMAIL THIS PAGE TO A FRIEND
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference
On-demand controlled release of anti-inflammatory and analgesic drugs from conducting polymer films to aid in wound healing.
PMID 23366114
Abstract
An electronically-controlled drug delivery system (eDDS) for the on-demand release of anti-inflammatory, anti-microbial and analgesic agents to aid in wound healing is currently under development. The loading of several drugs into conductive polymer films and their subsequent on-demand, controlled release upon application of an electrical potential to the polymer film has been demonstrated. The loading and release (active and passive) of Ibuprofen sodium salt - a negatively charged analgesic and anti-inflammatory agent - from polypyrrole films is described. Major challenges identified include precise control over drug loading and passive release from the conducting polymers in the absence of an applied potential.