EMAIL THIS PAGE TO A FRIEND
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference
Intradermal needle-free powdered drug injection by a helium-powered device.
PMID 23366327
Abstract
We present a new method for needle-free powdered drug injection via a bench-top gas-powered device. This injector provides an alternative method of vaccine delivery to address the cold chain problem--the cost and risk of transporting temperature sensitive vaccines to developing countries. The device houses interchangeable nozzle inserts to vary orifice geometries and is capable of delivering polymer beads (1-5 µm diameter) into the dermal layer of porcine tissue. Results for injection shape and injection depth versus nozzle orifice diameter demonstrate the device's controllability.