EMAIL THIS PAGE TO A FRIEND
Revue medicale suisse
[Pharmacovigilance update].
PMID 23367709
Abstract
Main pharmacovigilance updates in 2012 are reviewed here. Dabigatran: elderly patients with renal failure are at higher risk of bleeding. Dual renin-angiotensin-aldosterone system blockade comprising aliskiren is harmful. Incretins: low risk of acute pancreatitis. Interaction between fusidic acid and statins: many reports of rhabdomyolysis. Interactions between boceprevir/telaprevir and antiretroviral therapies: complex, but manageable. Citalopram, ondansetron: maximum recommended doses are reduced. Atomoxetine: significant increase in blood pressure and heart rate in a fraction of exposed patients. Agomelatine: elevated liver enzymes are common. Fingolimod: bradycardia and heart blocks after first dose - stronger safety recommendations regarding use in patients with heart conditions and strengthened cardiovascular monitoring.