EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Adsorption of lignocelluloses of model pre-hydrolysis liquor on activated carbon.
PMID 23370214
Abstract
The main objective of this work was to study the adsorption behavior of various components dissolved in the pre-hydrolysis of kraft process on activated carbon. In this work, model prehydrolysis liquor (PHL) solutions (MPHL)s were prepared via mixing various commercially available monosugars, xylan, lignin and furfural; and their adsorption performance on activated carbon (AC) was investigated. In singular (one component) MPHL/AC systems, furfural had the maximum and xylose had the minimum adsorption, and the adsorption of monosugars was basically similar on AC. Also, polydiallyldimethylammonium chloride (PDADMAC) was added (0.5 g/l) to singular xylan or lignin MPHL/AC system, which increased the lignin and xylan adsorptions to 350 and 190 mg/g on AC, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
471003
Lignin, alkali, low sulfonate content
370959
Lignin, alkali