EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Dissolution testing of amorphous solid dispersions.
PMID 23370435
Abstract
The main purpose of this study was to investigate the effect of different polymers, with varying physicochemical properties and molecular weight on the stability and dissolution of co-milled amorphous solid dispersions (ASDs) of piroxicam (PRX). The stability of amorphous PRX (aPRX) in ASDs was significantly improved by the polymers. In-line Raman spectroscopy revealed that solvent mediated solid state changes occurred in biorelevant medium, however differences between ASDs were found. Thus, the dissolution behavior of ASDs of PRX and the respective polymer during conventional large volume (900ml) and a commercial small volume (20ml) dissolution testing was evaluated. The results of these studies indicated that the molecular weight of the polymer (PVP90 vs PVP25) is influencing the solubility of PRX from ASD. Interestingly the effect of molecular weight of the polymer was different than reported previously in the literature for the similar ASDs prepared by spray drying. Furthermore, the dose related bioavailability was determined by investigating the experimental saturation concentrations for different doses. These studies confirmed the findings of the dissolution studies. The differences are presumably caused by the formation of physically different diffusion layers around the ASD particles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
02286
Kollidon® 25
64655
Methocel® 60 HG, 28-30% methoxyl basis
64670
Methocel® 65 HG, 27-29% methoxyl basis
64680
Methocel® 90 HG, 22-27% methoxyl basis
64605
Methocel® A15 LV, 27.5-31.5% methoxyl basis
64625
Methocel® A15C, medium viscosity, Methoxyl content 27.5-31.5 %
94378
Methocel® A4M, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
M0262
Methyl cellulose, viscosity: 400 cP
M7140
Methyl cellulose, viscosity: 15 cP
M6385
Methyl cellulose, viscosity: 25 cP
M7027
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
M0387
Methyl cellulose, viscosity: 1,500 cP
M0512
Methyl cellulose, viscosity: 4,000 cP
M0430
Methyl cellulose, meets USP testing specifications, viscosity: 400 cP
M0555
Methyl cellulose, meets USP testing specifications, viscosity: 1,500 cP
64632
Methyl cellulose, tested according to Ph.Eur.
P5654
Piroxicam, ≥98% (TLC)
C15H13N3O4S
P0847
Piroxicam, meets USP testing specifications
C15H13N3O4S
P2130000
Piroxicam, European Pharmacopoeia (EP) Reference Standard
C15H13N3O4S
Y0001475
Piroxicam for system suitability
C15H13N3O4S
PVP10
Polyvinylpyrrolidone, average mol wt 10,000
P2307
Polyvinylpyrrolidone, plant cell culture tested, average mol wt 10,000
PVP40
Polyvinylpyrrolidone, average mol wt 40,000
P0930
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo
PVP360
Polyvinylpyrrolidone, average mol wt 360,000
P5288
Polyvinylpyrrolidone, for molecular biology, nucleic acid hybridization tested, mol wt 360,000
90268
Polyvinylpyrrolidone, K 25, tested according to Ph.Eur.
81420
Polyvinylpyrrolidone, K 30
81440
Polyvinylpyrrolidone, K 90
234257
Polyvinylpyrrolidone, powder, average Mw ~29,000