EMAIL THIS PAGE TO A FRIEND
Journal of nuclear medicine technology
Post-90Y radioembolization PET/CT scan with respiratory gating using time-of-flight reconstruction.
PMID 23370544
Abstract
We present an interesting case of a post-(90)Y radioembolization PET scan acquired with the aide of respiratory gating. We postulated that respiratory gating would allow accurate tracking of the tumor position, which may be altered by patient breathing. Time-of-flight image reconstruction was used to produce images with less noise in fewer iterations than is possible with conventional PET.