EMAIL THIS PAGE TO A FRIEND
Dental Materials Journal
The effect of silica-coating by sol-gel process on resin-zirconia bonding.
PMID 23370886
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
221228
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
H5NO