EMAIL THIS PAGE TO A FRIEND
Applied biochemistry and biotechnology
Multistep conversion of para-substituted phenols by phenol hydroxylase and 2,3-dihydroxybiphenyl 1,2-dioxygenase.
PMID 23371781
Abstract
A multistep conversion system of para-substituted phenols by recombinant phenol hydroxylase (PH(IND)) and 2,3-dihydroxybiphenyl 1,2-dioxygenase (BphC(LA-4)) was constructed in this study. Docking studies with different para-substituted phenols and corresponding catechols inside of the active site of PH(IND) and BphC(LA-4) predicted that all the substrates should be transformed. High-performance liquid chromatography-mass spectrometry analysis showed that the products of multistep conversion were the corresponding para-substituted catechols and semialdehydes. For the first-step conversion, the formation rate of 4-fluorocatechol (0.39 μM/min/mg dry weight) by strain PH(IND) hydroxylation was 1.15, 6.50, 3.00, and 1.18-fold higher than the formation of 4-chlorocatechol, 4-bromocatechol, 4-nitrocatechol, and 4-methylcatechol, respectively. For the second-step conversion, the formation rates of semialdehydes by strain BphC(LA-4) were as follows: 5-fluoro-HODA>5-chloro-HODA>2-hydroxy-5-nitro-ODA>5-bromo-HODA>2-hydroxy-5-methyl-ODA. The present study suggested that the multistep conversion by both ring hydroxylase and cleavage dioxygenase should be potential in the synthesis of industrial precursors and provide a novel avenue in the wastewater recycling treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
148520
4-Chlorocatechol, 97%
C6H5ClO2