EMAIL THIS PAGE TO A FRIEND
Prescrire international
Estradiol + nomegestrol. Oral contraceptive claimed to be more natural, but presenting no obvious advantages.
PMID 23373095
Abstract
No evidence of an advantage over standard oestrogen-progestin combinations, but as many serious adverse effects as with ethinylestradiol + drospirenone.