EMAIL THIS PAGE TO A FRIEND
Managed care (Langhorne, Pa.)
Injectable drug may help avoid eye surgery in patients with vitreomacular adhesion.
PMID 23373143
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1867
Plasmin from human plasma, lyophilized powder, ≥2.0 units/mg protein