EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Effect of ginsenoside Rg1 on functional expression of human neural stem cells: a patch clamp study].
PMID 23373225
Abstract
To observe the effects of ginsenoside Rg1 on the functional expression of human neural stem cells (hNSCs). The membrane electrophysiological properties and sodium and potassium ion channels in the hNSCs induced by Rg1 were analyzed using the whole-cell patch-clamp. On the 7th day, the neuron-like cells derived from ginsenoside Rg1 (20 mg x L(-1))-induced NSCs show: (1) The resting membrane potential: (-45.70 +/- 2.63) mV, the membrane capacitance: (26.89 +/- 1.91) pF, the membrane input impedance: (877.51 +/- 20.44) MH (P < 0.05 compared with the control group, respectively); (2) The detection rate of inward sodium current which is rapidly activated and inactivated in voltage-dependence was 50%, and its average peak value was (711.48 +/- 158.03) pA (P < 0.05 compared with the control group); (3) The outward potassium currents were composed of rapidly activated and inactivated transient outward potassium current and delayed rectifier outward potassium current, and its average peak value was (1 070.42 +/- 177.18) pA (P < 0.05 compared with the control group). Ginsenoside Rg1 can promote the functional expression and maturity of hNSCs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
68317
Ginsenoside Rg1, analytical standard
C42H72O14
00370580
Ginsenoside Rg1, primary reference standard
C42H72O14