EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Retention models and interaction mechanisms of acetone and other carbonyl-containing molecules with amylose tris[(S)-?-methylbenzylcarbamate] sorbent.
PMID 23374367
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0512
Amylose from potato, used as amylase substrate