EMAIL THIS PAGE TO A FRIEND
Molecular genetics and metabolism
Pitfalls in phenylalanine loading test in the diagnosis of dopa-responsive dystonia.
PMID 23375473
Abstract
Phenylalanine (Phe) loading test is a useful tool in the differential diagnosis of dopa-responsive dystonia due to autosomal dominant or recessive GTP cyclohydrolase I (GTPCH) deficiency or autosomal recessive sepiapterin reductase (SR) deficiency. In these patients hepatic phenylalanine hydroxylase system is compromised due to subnormal tetrahydrobiopterin (BH(4)) levels and hydroxylation of phenylalanine (Phe) to tyrosine (Tyr) is reduced with elevated Phe/Tyr ratio 1-2 h after oral Phe administration (100 mg/kg bw) administration. In healthy persons there is only a modest increase in Tyr production and blood Phe normalizes after 4 h. We report on a challenge with Phe (100 mg/kg bw) in a patient with dopa-responsive dystonia while on therapy with BH(4) and l-dopa. During Phe challenge Phe concentration remained below the normal range while a transient mild hypertyrosinemia was observed, leading to an extremely low Phe/Tyr ratio. A repeated test, after BH(4) withdrawal, reversed the findings and resulted normal. These data suggest activation of hepatic phenylalanine hydroxylase by BH(4). Thus, the Phe loading test should not be performed during substitution with BH(4).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B2517
6-Biopterin, ≥97%
C9H11N5O3