EMAIL THIS PAGE TO A FRIEND
FEBS letters
Neohesperidin dihydrochalcone: presentation of a small molecule activator of mammalian alpha-amylase as an allosteric effector.
PMID 23376024
Abstract
Flavonoids and their precursor trans-chalcone have been reported as inhibitors of mammalian alpha-amylase. With regard to this background, neohesperidin dihydrochalcone (NHDC) effect was investigated toward porcine pancreatic alpha-amylase (PPA), and found to be an activator of the enzyme. The maximal activation (up to threefold) was found to occur at 4.8mM of NHDC, which could be considered to have a high activation profile, with regard to the alpha and beta parameters (alpha<1
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8220
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
10065 α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
86250 α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
A9857
α-Amylase from Aspergillus oryzae, powder, ≥150 units/mg protein (biuret)
A7595
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
A3403
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
A4582 α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
A4551
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
10067 α-Amylase from Bacillus licheniformis, thermostable amylase, powder, light brown, ≥2 U/mg
A4862 α-Amylase from Bacillus licheniformis, liquid
A6255
α-Amylase from porcine pancreas, DFP Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
A4268
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
50162
Hesperidin, analytical standard
C28H34O15
75041
Neohesperidin dihydrochalcone, analytical standard
C28H36O15
N8757
Neohesperidin dihydrochalcone, ≥95% (HPLC)
C28H36O15
W381101
Neohesperidin dihydrochalcone, ≥96%, FG
C28H36O15
86247 Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg