EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Growth, spectroscopic investigation, optical, mechanical, chemical etching and nonlinear optical studies of semi-organic crystal: Bis (thiourea) zinc formate.
PMID 23376259
Abstract
Single crystals of Bis (thiourea) zinc formate (BTZF) were grown by slow evaporation method at room temperature. The grown crystal was confirmed by single crystal XRD, (1)H NMR and Mass spectroscopic techniques. Single crystal XRD technique revealed that material crystallized in monoclinic system with cell parameters a=8.693Å, b=7.151Å and c=9.311Å. The presence of hydrogen atoms in the grown sample was confirmed by proton NMR analysis. The mass spectral analysis was carried out to measure the accurate molecular mass of the compound. The recorded UV-Vis-NIR transmittance spectrum show excellent transmission in the range of 200-1100nm. Measuring transmittance of BTZF permitted the calculation of the extinction coefficient K, Reflectance R, as functions of photon energy. The etching study indicates the occurrence of different types of etch pit patterns on the growth surface. The Vickers (Hv) microhardness were carried out in the load range of 10-50g. Optical nonlinearities of BTZF have been investigated by Z-scan technique with He-Ne laser radiation of wavelength 638nm.