EMAIL THIS PAGE TO A FRIEND
Gene
Involvement of a novel intronic microRNA in cross regulation of N-methyltransferase genes involved in caffeine biosynthesis in Coffea canephora.
PMID 23376454
Abstract
There are numerous reports on intronic miRNAs from plants, most of which are involved in the regulation of unrelated genes. Some of the target genes are antagonistic to the host genes. Intronic miRNAs in animal systems, however, are known to have synergistic effects. This article is the first to report a similar regulatory effect of a miRNA originating from an intron in plants. NMT genes involved in caffeine biosynthesis were silenced to obtain transformants with reduced caffeine. Transcript analysis revealed the accumulation of transcripts for a related NMT gene (CaMTL1) in transformants bearing either antisense or RNAi constructs. The altered expression was assumed to relate to the silencing of the NMT genes. Bioinformatics analysis of the genes involved in biosynthesis revealed the presence of an intronic miRNA originating from the intron of the theobromine synthase gene targeting CaMTL1. The putative miRNA was cloned and sequenced. Modified 5'-RLM-RACE mapping of the cleavage site and subsequent Northern blotting experimentally demonstrated the presence and activity of such a miRNA in Coffea canephora. This novel regulatory mechanism previously unreported in plants will shed more light onto the evolution of multigene families and the role of introns in this process.
Related Materials