EMAIL THIS PAGE TO A FRIEND
Nuclear medicine communications
Diagnostic role of 18F-dihydroxyphenylalanine positron emission tomography in patients with congenital hyperinsulinism: a meta-analysis.
PMID 23376859
Abstract
Studies have reported the applications of F-dihydroxyphenylalanine (F-DOPA) PET in patients with congenital hyperinsulinism (CHI). The aim of this study was to systematically review and perform a meta-analysis of published data on the diagnostic role of F-DOPA PET in patients with CHI. A comprehensive computer literature search of studies on F-DOPA PET or PET/computed tomography (CT) in patients with CHI was conducted. The pooled sensitivity and specificity of F-DOPA PET or PET/CT in patients with CHI were calculated. The area under the receiver-operating characteristic curve was calculated to measure the accuracy of F-DOPA PET or PET/CT in patients with CHI. Ten studies comprising 181 patients with CHI were included in this meta-analysis. The pooled sensitivity of F-DOPA PET and PET/CT in detecting CHI was 88% on a per-patient-based analysis. The pooled specificity of F-DOPA PET and PET/CT in demonstrating CHI was 79%. The area under the receiver-operating characteristic curve was 0.92 on a per-patient-based analysis. In patients with CHI, F-DOPA PET or PET/CT demonstrated high sensitivity and specificity. F-DOPA PET and PET/CT are accurate methods for the diagnosis of CHI. Nevertheless, possible sources of false-positive and false-negative results should be kept in mind.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9503
3,4-Dihydroxy-DL-phenylalanine
C9H11NO4