EMAIL THIS PAGE TO A FRIEND
Annals of clinical psychiatry : official journal of the American Academy of Clinical Psychiatrists
Two cases of delirium with agomelatine therapy.
PMID 23376872
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1362
Agomelatine, ≥98% (HPLC)
C15H17NO2