EMAIL THIS PAGE TO A FRIEND
Journal of drugs in dermatology : JDD
A split-faced, observer-blinded comparison study of topical adapalene/benzoyl peroxide and adapalene in the treatment of Asian acne patients.
PMID 23377384