EMAIL THIS PAGE TO A FRIEND
Journal of drugs in dermatology : JDD
A double-blind, randomized, bilateral comparison of skin irritancy following application of the combination acne products clindamycin/tretinoin and benzoyl peroxide/adapalene.
PMID 23377511
Abstract
The use of topical medications for acne vulgaris is often limited by their irritant properties. Newer combination preparations are available and offer convenience, but irritant potential may still be a hindrance, perhaps more so with the combination of 2 agents. Few studies have compared these formulations directly for tolerability. We sought to compare the tolerability of 2 combination topical acne products, clindamycin 1.2%-tretinoin 0.025% (CLIN/RA) gel and benzoyl peroxide 2.5%-adapalene 0.1% (BPO/ADA) gel. CLIN/RA and BPO/ADA were applied daily to opposite sides of a subject's face for 21 days in a double-blinded fashion. Investigators' Global Assessments and study subject self-assessments of burning/stinging, itching, erythema, and dryness/scaling were collected. Transepidermal water loss (TEWL) was also measured as an objective measure of skin irritation. A mixed model analysis and repeated-measures analysis of variance were used to compare outcomes for both acne formulations. CLIN/RA produced significantly less burning/stinging than BPO/ADA (P<.001) as well as significantly less pruritus than BPO/ ADA (P<.001). BPO/ADA caused significantly more TEWL than CLIN/RA (P=.005). There was no significant difference in the amount of erythema or the amount of dryness/scaling caused by either formulation. CLIN/RA produced significantly less skin irritancy and TEWL than BPO/ADA.