EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biochemistry
AdipoR1 and AdipoR2 gene expression are regulated by thyroid hormones in adipose tissue.
PMID 23378066
Abstract
The aim of this study was to examine whether the relative gene expression of AdipoR1 and AdipoR2 in rat adipose tissue is altered by thyroid hormones, and whether this might relate to their circulating thyroid hormones and adiponectin levels. Hyper- and hypothyroidism were induced by daily oral administration of levothyroxine and methimazole in rats, respectively, over a 42 days period. Real-time PCR analysis was performed to evaluate the changes in AdipoR1 and AdipoR2 mRNA levels in the adipose tissue on days 15, 28, 42, and also 2 weeks after the cessation of treatment. In response to treatment with methimazole, mRNA levels of AdipoR1 and AdipoR2 decreased in the white adipose tissue compared to the euthyroid rats (p < 0.05). This decline was reversible 2 weeks after treatment cessation. The mRNA levels of AdipoR1 and AdipoR2 were increased in the hyperthyroid group of animals compared to euthyroid control (p < 0.05), and its changes were reversible 2 weeks after treatment cessation (P < 0.05). Adiponectin receptors gene expression levels in the adipose tissue of treated animals have positive correlations with thyroid hormones concentrations. Our results suggest that AdipoR1 and AdipoR2 gene expression is regulated by thyroid hormones in hypo- and hyperthyroidism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46429
Methimazole, VETRANAL, analytical standard
C4H6N2S