EMAIL THIS PAGE TO A FRIEND
Journal of radiology case reports
Children presenting with calcified disc herniation: a self-limiting process.
PMID 23378872
Abstract
We present two cases of disc herniation associated with juvenile intervertebral disc calcification, a rare, self-limiting process which typically resolves with conservative treatment. Recognition of this entity prevents unnecessary diagnostic workup and possible surgical intervention. A review of the literature for this rare entity is discussed.