EMAIL THIS PAGE TO A FRIEND
Perspective infirmiere : revue officielle de l'Ordre des infirmieres et infirmiers du Quebec
[Xarelto: other advantages].
PMID 23379053
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F9302
Factor X Activated (Xa) from bovine plasma, aqueous glycerol solution