EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Toxicological evaluation of pure hydroxytyrosol.
PMID 23380205
Abstract
Of all the phenolic constituents of olives and extra virgin olive oil, hydroxytyrosol is currently being actively exploited as a potential supplement or preservative to be employed in the nutraceutical, cosmeceutical, and food industry. In terms of safety profile, hydroxytyrosol has only been investigated as the predominant part of raw olive mill waste water extracts, due to the previous unavailability of appropriate quantities of the pure compound. We report the toxicological evaluation of hydroxytyrosol and, based on the results, propose a No Observed Adverse Effects Level (NOAEL) of 500mg/kg/d.