EMAIL THIS PAGE TO A FRIEND
Biorheology
Effects of long-term feeding of α-glucosylhesperidin on the mechanical properties of rabbit femoral arteries.
PMID 23380901
Abstract
Many people are sensitive to cold, resulting in poor blood circulation. There is evidence that hesperidin results in increased peripheral circulation and skin temperature. A transglycosylated hesperidin, α-glucosylhesperidin, is more bioabsorbable than hesperidin. In the present study, biomechanical studies were performed on the effects of long-term feeding of α-glucosylhesperidin on the contractile response (diameter response) and stiffness of femoral arteries excised from rabbits. Animals in the normal (non-treated), low, and high groups were fed 0, 150 and 4500 mg/day, respectively, of α-glucosylhesperidin for about 24 weeks. The feeding of α-glucosylhesperidin did not change arterial stiffness nor mean blood flow rate in the femoral artery; however, it increased mean aortic blood pressure and decreased arterial diameter at 100 mmHg in the high group. The diameter responses developed by 10-5 M of norepinephrine were significantly lower in the high and low groups than in non-treated group. This result indicates that, due to the long-term feeding of α-glucosylhesperidin, arterial contraction induced by the neurotransmitter of sympathetic nerves decreases. It was estimated that blood flow in such muscular arteries as the femoral artery is maintained at normal by α-glucosylhesperidin even under the conditions of autonomic imbalance and cold intolerance.