EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Biohybrid -Se-S- coupling reactions of an amino acid derived seleninate.
PMID 23381022
Abstract
We describe the synthesis of the N-(2-seleninatoethyl) amide of N-Boc-phenylalanine, serving here as a peptide model, and its reductive coupling reactions under mild conditions with unprotected thiouridine and glutathione. Selenosulfide products such as these comprise reversibly conjugated bio-components, and can potentially find uses as probes of biological function, such as enzyme inhibitors, delivery systems, or structural mimics.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T4509
4-Thiouridine, ≥98%
C9H12N2O5S