EMAIL THIS PAGE TO A FRIEND
The Medical letter on drugs and therapeutics
Apixaban (Eliquis)--a new oral anticoagulant for atrial fibrillation.
PMID 23381226
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F9302
Factor X Activated (Xa) from bovine plasma, aqueous glycerol solution