PloS one
Regulation of two renal chloride transporters, AE1 and pendrin, by electrolytes and aldosterone.
PMID 23383138
Abstract
The renal handling of salt and protons and bicarbonate are intricately linked through shared transport mechanisms for sodium, chloride, protons, and bicarbonate. In the collecting duct, the regulated fine-tuning of salt and acid-base homeostasis is achieved by a series of transport proteins located in different cell types, intercalated and principal cells. Intercalated cells are considered to be of less importance for salt handling but recent evidence has suggested that the anion exchanger pendrin may participate in salt reabsorption and blood pressure regulation. Here, we examined the regulated expression of two functionally related but differentially expressed anion exchangers, AE1 and pendrin, by dietary electrolyte intake and aldosterone. Cortical expression of pendrin was regulated on mRNA and protein level. The combination of NaHCO₃ and DOCA enhanced pendrin mRNA and protein levels, whereas DOCA or NaHCO₃ alone had no effect. NaCl or KHCO₃ increased pendrin mRNA, KCl decreased its mRNA abundance. On protein level, NH₄Cl, NaCl, and KCl reduced pendrin expression, the other treatments were without effect. In contrast, AE1 mRNA or protein expression in kidney cortex was regulated by none of these treatments. In kidney medulla, NaHCO₃/DOCA or NaHCO₃ alone enhanced AE1 mRNA levels. AE1 protein abundance was increased by NH₄Cl, NaHCO₃/DOCA, and NaCl. Immunolocalization showed that during NH₄Cl treatment the relative number of AE1 positive cells was increased and pendrin expressing cells reduced. Thus, pendrin and AE1 are differentially regulated with distinct mechanisms that separately affect mRNA and protein levels. Pendrin is regulated by acidosis and chloride intake, whereas AE1 is enhanced by acidosis, NaCl, and the combination of DOCA and NaHCO₃.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4514
Ammonium chloride, ReagentPlus®, ≥99.5%
H4ClN
11209
Ammonium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)
H4ClN
431583
Potassium bicarbonate, ≥99.95% trace metals basis, 99.7-100.5% dry basis
CHKO3
237205
Potassium bicarbonate, ACS reagent, 99.7%, powder, crystals or granules
CHKO3
60339
Potassium bicarbonate, BioUltra, ≥99.5% (T)
CHKO3
60340
Potassium bicarbonate, purum p.a., ≥99.0% (T), powder, crystals or chunks
CHKO3
12602
Potassium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, E501, 99.5-101.0% (acidimetric)
CHKO3
S6014
Sodium bicarbonate, ACS reagent, ≥99.7%
CHNaO3
S8875
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
CHNaO3
31437
Sodium bicarbonate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.7%, crystals
CHNaO3
13433
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
CHNaO3
900506
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
CHNaO3
36486
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
S8761
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
401676
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
CHNaO3