PloS one
COMT and ANKK1-Taq-Ia genetic polymorphisms influence visual working memory.
PMID 23383291
Abstract
Complex cognitive tasks such as visual working memory (WM) involve networks of interacting brain regions. Several neurotransmitters, including an appropriate dopamine concentration, are important for WM performance. A number of gene polymorphisms are associated with individual differences in cognitive task performance. COMT, for example, encodes catechol-o-methyl transferase the enzyme primarily responsible for catabolizing dopamine in the prefrontal cortex. Striatal dopamine function, linked with cognitive tasks as well as habit learning, is influenced by the Taq-Ia polymorphism of the DRD2/ANKK1 gene complex; this gene influences the density of dopamine receptors in the striatum. Here, we investigated the effects of these polymorphisms on a WM task requiring the maintenance of 4 or 6 items over delay durations of 1 or 5 seconds. We explored main effects and interactions between the COMT and DRD2/ANKK1-Taq-Ia polymorphisms on WM performance. Participants were genotyped for COMT (Val(158)Met) and DRD2/ANKK1-Taq-Ia (A1+, A1-) polymorphisms. There was a significant main effect of both polymorphisms. Participants' WM reaction times slowed with increased Val loading such that the Val/Val homozygotes made the slowest responses and the Met/Met homozygotes were the fastest. Similarly, WM reaction times were slower and more variable for the DRD2/ANKK1-Taq-Ia A1+ group than the A1- group. The main effect of COMT was only apparent in the DRD2/ANKK1-Taq-Ia A1- group. These findings link WM performance with slower dopaminergic metabolism in the prefrontal cortex as well as a greater density of dopamine receptors in the striatum.