EMAIL THIS PAGE TO A FRIEND
Pain medicine (Malden, Mass.)
Prolonged delirium with psychotic features from omega conotoxin toxicity.
PMID 23383992
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1182
ω-Conotoxin MVIIA, ≥95% (HPLC)
C102H172N36O32S7