EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Determination of monosaccharides derivatized with 2-aminobenzoic Acid by capillary electrophoresis.
PMID 23386335
Abstract
Reducing monosaccharides were derivatized with 2-aminobenzoic acid (2-AA) through reductive amination using sodium cyanoborohydride as a reductant, and the derivatives were separated by capillary zone electrophoresis with UV detection using 50 mM sodium phosphate (pH 5.5) or 150 mM sodium borate-50 mM sodium phosphate (pH 7.0) running buffer. The derivatives of monosaccharides, which are major components of various carbohydrate materials, were completely separated within 25 min.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
10680
Anthranilic acid, puriss. p.a., ≥99.5% (T)
C7H7NO2
A89855
Anthranilic acid, reagent grade, ≥98%
C7H7NO2
71997
Borax Anhydrous, BioUltra, anhydrous, ≥99%
B4Na2O7
72001
Borax Anhydrous, anhydrous, decomposing agent (for x-ray fluorescence analysis), ≥99%
B4Na2O7
71996
Borax Anhydrous, puriss. p.a., anhydrous, ≥98.0% (T)
B4Na2O7
S9763
Sodium phosphate dibasic, ACS reagent, ≥99.0%
HNa2O4P
71640
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
HNa2O4P
04276
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
HNa2O4P
71650
Sodium phosphate dibasic dodecahydrate, puriss. p.a., crystallized, ≥99.0% (T)
HNa2O4P · 12H2O
04273
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-101% (calc. to the dried substance)
HNa2O4P · 12H2O
431478
Sodium phosphate dibasic heptahydrate, ≥99.99% trace metals basis
HNa2O4P · 7H2O
S9390
Sodium phosphate dibasic heptahydrate, ACS reagent, 98.0-102.0%
HNa2O4P · 7H2O
S2429
Sodium phosphate dibasic heptahydrate, meets USP testing specifications
HNa2O4P · 7H2O
71911
Sodium phosphate tribasic dodecahydrate, puriss. p.a., ACS reagent, ≥98.0% (T)
Na3O4P · 12H2O
222003
Sodium phosphate tribasic dodecahydrate, ACS reagent, ≥98%
Na3O4P · 12H2O
229946
Sodium tetraborate, 99.998% trace metals basis
B4Na2O7
221732
Sodium tetraborate, 99%
B4Na2O7