EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Palladium-catalyzed amination of dichloroquinolines with adamantane-containing amines.
PMID 23389254
Abstract
Pd-catalyzed amination of isomeric 2,6-, 2,8-, 4,8- and 4,7-dichloroquinolines was studied using adamantane-containing amines in which substituents at the nitrogen atom differ in bulkiness. The selectivity of the amination of 2,6-dichloroquinoline was very low, substantially better results were obtained with 2,8-dichloroquinoline, and 4,8- and 4,7-dichloroquinolines provided the best yields of the amination products. Diamination of 4,8- and 4,7-dichloroquinolines was carried out with two amines which differ strongly in the bulkiness of the alkyl group. In the majority of cases BINAP ligand was successfully applied, however, it had to be replaced with DavePhos in certain reactions when using the most sterically hindered amine as well as for the diamination reactions.