EMAIL THIS PAGE TO A FRIEND
Optics express
Optimal overlayer inspired by Photuris firefly improves light-extraction efficiency of existing light-emitting diodes.
PMID 23389270
Abstract
In this paper the design, fabrication and characterization of a bioinspired overlayer deposited on a GaN LED is described. The purpose of this overlayer is to improve light extraction into air from the diode's high refractive-index active material. The layer design is inspired by the microstructure found in the firefly Photuris sp. The actual dimensions and material composition have been optimized to take into account the high refractive index of the GaN diode stack. This two-dimensional pattern contrasts other designs by its unusual profile, its larger dimensions and the fact that it can be tailored to an existing diode design rather than requiring a complete redesign of the diode geometry. The gain of light extraction reaches values up to 55% with respect to the reference unprocessed LED.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
57083
Indium, bars, 99.95%
In
264113
Indium, beads, diam. 2-5 mm, 99.999% trace metals basis
In
326607
Indium, beads, diam. 2-5 mm, ≥99.9% trace metals basis
In
357278
Indium, foil, thickness 1.0 mm, 99.999% trace metals basis
In
326631
Indium, foil, thickness 0.5 mm, 99.999% trace metals basis
In
357286
Indium, foil, thickness 0.5 mm, 99.99% trace metals basis
In
264040
Indium, foil, thickness 0.25 mm, 99.995% trace metals basis
In
357294
Indium, foil, thickness 0.25 mm, 99.99% trace metals basis
In
264059
Indium, foil, thickness 0.127 mm, 99.99% trace metals basis
In
357308
Indium, foil, thickness 0.1 mm, ≥99.995% trace metals basis
In
326615
Indium, pieces, 99.99% trace metals basis
In
203432
Indium, powder, −60 mesh, 99.999% trace metals basis
In
264032
Indium, powder, −100 mesh, 99.99% trace metals basis
In
277959
Indium, powder, 99.99% trace metals basis
In
385379
Indium, rod, diam. 10 mm, 99.9998% trace metals basis
In
264091
Indium, rod, diam. 6 mm, ≥99.999% trace metals basis
In
357065
Indium, wire, diam. 2.0 mm, 99.995% trace metals basis
In
340863
Indium, wire, diam. 1.5 mm, 99.99% trace metals basis
In
357073
Indium, wire, diam. 1.0 mm, 99.995% trace metals basis
In
264075
Indium, wire, diam. 1.0 mm, 99.99% trace metals basis
In
264067
Indium, wire, diam. 0.5 mm, 99.995% trace metals basis
In
326623
Indium, wire, diam. 0.25 mm, 99.999% trace metals basis
In