EMAIL THIS PAGE TO A FRIEND
Endocrinology and metabolism clinics of North America
Central leucine sensing in the control of energy homeostasis.
PMID 23391241
Abstract
This article reviews recent studies identifying two key brain regions as two critical nodes in the neural network where central leucine sensing contributes to whole body energy homeostasis: the mediobasal hypothalamus and the dorsal vagal complex of the caudal brainstem. Activation of these leucine sensing sites engages multiple determinants of energy balance, including glucose homeostasis, food intake, and adiposity.