EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Isolation of the putative biosynthetic gene cluster of 1-deoxynojirimycin by Bacillus amyloliquefaciens 140N, its production and application to the fermentation of soybean paste.
PMID 23391926
Abstract
The 1-deoxynojirimycin (DNJ) biosynthetic gene cluster of Bacillus amyloliquefaciens 140N isolated from traditional Korean fermented food was isolated by PCR screening. It showed 78.9% inhibitory activity against α-glucosidase and produced 0.8 g/L of DNJ in an optimized medium containing 2% soluble starch, 1% tryptone, 0.05% KH(2)PO(4), and 0.05% (NH(2))(4)SO(4). Soybean paste fermented with B. amyloliquefaciens 140N produced DNJ with 84.4% inhibitory activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9305
1-Deoxynojirimycin hydrochloride
C6H13NO4 · HCl