EMAIL THIS PAGE TO A FRIEND
Digestive diseases and sciences
First report of telaprevir-induced pancreatitis.
PMID 23392743
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
62305 Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg