EMAIL THIS PAGE TO A FRIEND
Clinical drug investigation
Effect of add-on pentoxifylline on proteinuria in membranous glomerulonephritis: a 6-month placebo-controlled trial.
PMID 23392759
Abstract
Membranous glomerulonephritis (MGN) may cause proteinuria as the main complication and is a strong risk factor for end-stage renal disease. Current therapeutic regimens provide only partial renoprotection. Data derived from both animal and human studies provide a scientific basis for the use of pentoxifylline as an antiproteinuric agent. This study was designed to evaluate the antiproteinuric effect of add-on pentoxifylline therapy in non-diabetic patients with MGN. This was a double-blind, placebo-controlled trial. Non-diabetic patients with histologically proven MGN and urinary protein excretion (UPE) > 500 mg/24 h, entered a 6-month study period. Enrolled patients were selected from a university and three private clinics. Patients were assigned to one of the two treatment groups: pentoxifylline 400 mg two or three times a day, or matching placebo. Baseline and follow-up assessments included estimated glomerular filtration rate (eGFR) and UPE. Differences in the changes in variables within the placebo and pentoxifylline treatment groups during the study period were assessed using Friedman's test. Treatment with pentoxifylline for 6 months resulted in a significant reduction of mean UPE (p < 0.001) along with a slight, non-significant increase of eGFR, in comparison to the mean UPE and eGFR increase in the placebo group. This study showed that add-on therapy of pentoxifylline in MGN was beneficial, and could be considered as a potential new therapeutic indication for the drug in such kidney diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0510000
Pentoxifylline, European Pharmacopoeia (EP) Reference Standard
C13H18N4O3